Methyl 5-bromothiophene-3-carboxylate 97%

Supplier: Ambeed
Warning

A179184-250MG A179184-25G A179184-5G A179184-1G
76985-684EA 29.71 USD
76985-684 76985-686 76985-688 77683-434
Methyl 5-bromothiophene-3-carboxylate 97%
Methyl 5-bromothiophene-3-carboxylate

Precaución: For research use only.

Formula: C₆H₅BrO₂S
MW: 221.072 g/mol
MDL Number: MFCD13386488
CAS Number: 88770-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR