Isobutyl 5-chloro-2,2-dimethylvalerate 97%

Supplier: Ambeed
Warning

A179179-1G A179179-250MG A179179-5G
76998-578EA 25 USD
76998-578 76998-580 76998-582
Isobutyl 5-chloro-2,2-dimethylvalerate 97%
Isobutyl 5-chloro-2,2-dimethylvalerate

Precaución: for Research Use Only

Formula: C₁₁H₂₁ClO₂
MW: 220.74 g/mol
MDL Number: MFCD03265308
CAS Number: 109232-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR