(R)-2-(((2S,3S)-4-((E)-3-((R)-1-Carboxy-2-(3,4-dihydroxyphenyl)ethoxy)-3-oxoprop-1-en-1-yl)-2-(3,4-dihydroxyphenyl)-7-hydroxy-2,3-dihydrobenzofuran-3-carbonyl)oxy)-3-(3,4-dihydroxyphenyl)propanoic acid 98%

Supplier: Ambeed
Warning

A179169-100MG A179169-10MG A179169-1G A179169-1MG A179169-250MG A179169-25MG A179169-5G A179169-5MG
76998-562EA 121 USD
76998-562 76998-564 76998-566 76998-568 76998-570 76998-572 76998-574 76998-576
(R)-2-(((2S,3S)-4-((E)-3-((R)-1-Carboxy-2-(3,4-dihydroxyphenyl)ethoxy)-3-oxoprop-1-en-1-yl)-2-(3,4-dihydroxyphenyl)-7-hydroxy-2,3-dihydrobenzofuran-3-carbonyl)oxy)-3-(3,4-dihydroxyphenyl)propanoic acid 98%
(R)-2-(((2S,3S)-4-((E)-3-((R)-1-Carboxy-2-(3,4-dihydroxyphenyl)ethoxy)-3-oxoprop-1-en-1-yl)-2-(3,4-dihydroxyphenyl)-7-hydroxy-2,3-dihydrobenzofuran-3-carbonyl)oxy)-3-(3,4-dihydroxyphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₃₆H₃₀O₁₆
MDL Number: MFCD07779133
CAS Number: 121521-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR