1,3-Bis(4-aminophenoxy)benzene 98%

Supplier: Ambeed
Warning

A179107-100G A179107-25G A179107-500G
77029-358EA 24 USD
77029-358 77029-360 77029-362 77029-364
1,3-Bis(4-aminophenoxy)benzene 98%
1,3-Bis(4-aminophenoxy)benzene

Precaución: for Research Use Only

Formula: C₁₈H₁₆N₂O₂
MDL Number: MFCD00039154
CAS Number: 2479-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR