Boc-L-β-homoaspartic acid-γ-benzyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Boc-β-HomoAsp(OBzl)-OH, Boc-β-HoAsp(OBzl)-OH

A179098-10G A179098-1G A179098-250MG A179098-5G
76998-544EA 667 USD
76998-544 76998-546 76998-548 76998-550
Boc-L-β-homoaspartic acid-γ-benzyl ester 95%
Boc-L-beta-homoaspartic acid-gamma-benzyl ester

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₆
MDL Number: MFCD01862861
CAS Number: 254101-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR