3-Fluoro-2-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A179070-10G A179070-1G A179070-250MG A179070-25G A179070-5G
76998-534EA 63 USD
76998-534 76998-536 76998-538 76998-540 76998-542
3-Fluoro-2-methoxybenzoic acid 98%
3-Fluoro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MDL Number: MFCD00973972
CAS Number: 106428-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR