1-Phenylpiperidine-4-carboxamide 95%

Supplier: Ambeed
Warning

A179064-100MG A179064-1G A179064-250MG
77011-906EA 28.65 USD
77011-906 77011-908 77011-910
1-Phenylpiperidine-4-carboxamide 95%
1-Phenylpiperidine-4-carboxamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O
MW: 204.27 g/mol
MDL Number: MFCD08597043
CAS Number: 170353-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR