N-(4-Chloro-3-iodopyridin-2-yl)-2,2-dimethylpropionamide 98+%

Supplier: Ambeed
Warning

Synonyms: N-(4-Chloro-3-iodopyridin-2-yl)pivalamide

A179001-100MG A179001-1G A179001-250MG
77011-900EA 113.52 USD
77011-900 77011-902 77011-904
N-(4-Chloro-3-iodopyridin-2-yl)-2,2-dimethylpropionamide 98+%
N-(4-Chloro-3-iodopyridin-2-yl)-2,2-dimethylpropionamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClIN₂O
MW: 338.58 g/mol
MDL Number: MFCD08235131
CAS Number: 898561-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR