Methyl-2-methylthiophene-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-methylthiophene-3-carboxylate

A178984-10G A178984-1G A178984-25G A178984-5G
77029-350EA 263.11 USD
77029-350 77029-352 77029-354 77029-356
Methyl-2-methylthiophene-3-carboxylate 97%
Methyl-2-methylthiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈O₂S
MW: 156.2000 g/mol
MDL Number: MFCD12026360
CAS Number: 53562-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR