Boc-N-α-methyl-D-phenylalanine . DCHA 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-D-N-Me-Phe DCHA, Boc-D-N-Me-Phe-OH·DCHA
A178890-1G
A178890-5G
77029-346EA
13
USD
77029-346
77029-348
Boc-N-α-methyl-D-phenylalanine . DCHA 95%
Boc-N-alpha-methyl-D-phenylalanine . DCHA
Precaución: for Research Use Only
Formula:
C₁₅H₂₁NO₄·C₁₂H₂₃N MW: 460.66 g/mol |
MDL Number:
MFCD00058079 CAS Number: 102185-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...