2-Bromo-6-(bromomethyl)naphthalene 95%

Supplier: Ambeed
Danger

A178855-100MG A178855-1G
77029-342EA 29.71 USD
77029-342 77029-344
2-Bromo-6-(bromomethyl)naphthalene 95%
2-Bromo-6-(bromomethyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₁H₈Br₂
MW: 299.98 g/mol
MDL Number: MFCD09842471
CAS Number: 305798-02-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR