Methyl 6-methylbenzofuran-2-carboxylate 96%

Supplier: Ambeed
Warning

A178793-1G A178793-250MG
76985-678EA 82.76 USD
76985-678 76985-680
Methyl 6-methylbenzofuran-2-carboxylate 96%
Methyl 6-methylbenzofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₃
MW: 190.20 g/mol
MDL Number: MFCD11975072
CAS Number: 82788-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR