4-Methyl-1-indanone 97%

Supplier: Ambeed
Warning

A178752-1G A178752-250MG A178752-5G A178752-25G A178752-10G
76998-528EA 11 USD
76998-528 76998-530 76998-532 77567-634 77881-355
4-Methyl-1-indanone 97%
4-Methyl-1-indanone

Precaución: For research use only.

Formula: C₁₀H₁₀O
MDL Number: MFCD00015759
CAS Number: 24644-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR