3,6-Dibromo-4-methylpyridazine 98%

Supplier: Ambeed
Warning

A178741-1G A178741-250MG A178741-5G A178741-25G
77011-888EA 22 USD
77011-888 77011-890 77011-892 77890-141
3,6-Dibromo-4-methylpyridazine 98%
3,6-Dibromo-4-methylpyridazine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MDL Number: MFCD22056210
CAS Number: 89284-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR