2-(2-Aminoethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Pyridylethylamine, 2-(2-Pyridyl)ethylamine, 2-Pyridin-2-ylethanamine

A178734-100G A178734-1G A178734-25G A178734-5G
76998-520EA 143.23 USD
76998-520 76998-522 76998-524 76998-526
2-(2-Aminoethyl)pyridine 98%
2-(2-Aminoethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00006367
CAS Number: 2706-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR