1-Ethyl-5-methyl-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A178540-100MG A178540-250MG
76998-514EA 12.74 USD
76998-514 76998-516
1-Ethyl-5-methyl-1H-pyrazole-3-carboxylic acid 95%
1-Ethyl-5-methyl-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MDL Number: MFCD08445953
CAS Number: 50920-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR