FMOC-D-3,4-difluorophenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Phe(3,4-DiF)-OH

A178502-10G A178502-1G A178502-5G
76998-502EA 196 USD
76998-502 76998-504 76998-506
FMOC-D-3,4-difluorophenylalanine 95%
FMOC-D-3,4-difluorophenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₁₉F₂NO₄
MW: 423.42 g/mol
MDL Number: MFCD00672551
CAS Number: 198545-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR