1,5-Dimethyl-1H-pyrazol-3-ylamine 97%

Supplier: Ambeed
Warning

Synonyms: 1,5-Dimethyl-1H-pyrazol-3-amine, 3-Amino-1,5-dimethyl-1H-pyrazole

A178365-100MG A178365-10G A178365-1G A178365-250MG
76998-494EA 13.8 USD
76998-494 76998-496 76998-498 76998-500
1,5-Dimethyl-1H-pyrazol-3-ylamine 97%
1,5-Dimethyl-1H-pyrazol-3-ylamine

Precaución: for Research Use Only

Formula: C₅H₉N₃
MW: 111.15 g/mol
MDL Number: MFCD04970041
CAS Number: 35100-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR