8-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride 95%

Supplier: Ambeed
Warning

A178240-100MG A178240-250MG
77011-864EA 115 USD
77011-864 77011-866
8-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride 95%
8-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆Cl₂N₂
MDL Number: MFCD05664035
CAS Number: 17783-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR