Methyl-6-methoxypicolinate 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-pyridine-2-carboxylic acid methyl ester, Methyl 6-methoxypyridine-2-carboxylate, Methyl 6-methoxypicolinate

A178187-1G A178187-25G A178187-5G
77011-858EA 16 USD
77011-858 77011-860 77011-862
Methyl-6-methoxypicolinate 98%
Methyl 6-methoxypicolinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.1600 g/mol
MDL Number: MFCD07779517
CAS Number: 26256-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR