4-(2-Fluorobenzyl)piperidine hydrochloride 95%

Supplier: Ambeed
Warning

A178116-100MG A178116-1G
77011-856EA 16.98 USD
77011-856 77683-426
4-(2-Fluorobenzyl)piperidine hydrochloride 95%
4-(2-Fluorobenzyl)piperidine hydrochloride

Precaución: For research use only.

Formula: C₁₂H₁₆FN·ClH
MW: 229.721 g/mol
MDL Number: MFCD09787475
CAS Number: 193357-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR