Methyl 6-hydroxy-1H-indole-3-carboxylate 95%

Supplier: Ambeed
Warning

A178102-1G A178102-250MG A178102-5G
76985-662EA 131 USD
76985-662 76985-664 76985-666
Methyl 6-hydroxy-1H-indole-3-carboxylate 95%
Methyl 6-hydroxy-1H-indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MDL Number: MFCD11617113
CAS Number: 112332-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR