7-Hydroxy-3-(3-hydroxy-4-methoxyphenyl)-4H-chromen-4-one 98%

Supplier: Ambeed
Warning

Synonyms: Calycosin

A178055-10MG A178055-1MG A178055-5MG
77029-324EA 120.95 USD
77029-324 77029-326 77029-328
7-Hydroxy-3-(3-hydroxy-4-methoxyphenyl)-4H-chromen-4-one 98%
7-Hydroxy-3-(3-hydroxy-4-methoxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₅
MW: 284.26 g/mol
MDL Number: MFCD00210598
CAS Number: 20575-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR