5-Amino-3-methyl-1H-pyrazole-4-carbonitrile 98%

Supplier: Ambeed
Warning

A178043-100MG A178043-1G A178043-250MG A178043-25G A178043-5G
76998-470EA 16.98 USD
76998-470 76998-472 76998-474 76998-476 76998-478
5-Amino-3-methyl-1H-pyrazole-4-carbonitrile 98%
5-Amino-3-methyl-1H-pyrazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₆N₄
MDL Number: MFCD02020550
CAS Number: 5453-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR