(S)-1-(2-Chlorophenyl)ethanol 98%

Supplier: Ambeed
Warning

A177905-1G A177905-250MG A177905-5G
76998-464EA 23 USD
76998-464 76998-466 76998-468
(S)-1-(2-Chlorophenyl)ethanol 98%
(S)-1-(2-Chlorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MDL Number: MFCD06659506
CAS Number: 131864-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR