1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R,2S)-2-hydroxy-2,3-dihydro-1H-inden-1-yl)thiourea 97%
Supplier: Ambeed
|
Warning
|
A177826-100MG
A177826-5G
A177826-1G
A177826-250MG
77011-844EA
38.2
USD
77011-844
77683-424
77683-422
77683-423
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R,2S)-2-hydroxy-2,3-dihydro-1H-inden-1-yl)thiourea 97%
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R,2S)-2-hydroxy-2,3-dihydro-1H-inden-1-yl)thiourea
Precaución: For research use only.
Formula:
C₁₈H₁₄F₆N₂OS MW: 420.37 g/mol |
MDL Number:
MFCD27922602 CAS Number: 871828-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...