trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 97%

Supplier: Ambeed
Warning

A177808-10G A177808-1G A177808-250MG A177808-25G A177808-5G A177808-100G
76998-448EA 96 USD
76998-448 76998-450 76998-452 76998-454 76998-456 77881-350
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 97%
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₁F₃O₂
MDL Number: MFCD09037363
CAS Number: 133261-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR