2-(Boc-amino)-5-chloro-3-hydroxypyridine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (5-chloro-3-hydroxypyridin-2-yl)carbamate

A177802-100MG A177802-10G A177802-250MG
77011-838EA 14 USD
77011-838 77011-840 77011-842
2-(Boc-amino)-5-chloro-3-hydroxypyridine 97%
2-(Boc-amino)-5-chloro-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClN₂O₃
MW: 244.675 g/mol
MDL Number: MFCD27935389
CAS Number: 1609402-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR