(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-tert-Butyl L-Aspartate, H-Asp(OtBu)-OH, L-Aspartic acid 4-tert-butyl ester, L-Aspartic acid 4-tert-Butyl ester

A177744-100G A177744-10G A177744-1G A177744-25G A177744-5G
76998-428EA 173 USD
76998-428 76998-430 76998-432 76998-434 76998-436
(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid 98%
(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₅NO₄
MDL Number: MFCD00038577
CAS Number: 3057-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR