5-(Difluoromethyl)pyridin-3-amine 95%

Supplier: Ambeed
Danger

A177700-100MG A177700-1G
77011-834EA 135 USD
77011-834 77011-836
5-(Difluoromethyl)pyridin-3-amine 95%
5-(Difluoromethyl)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₆F₂N₂
MDL Number: MFCD18384496
CAS Number: 899898-94-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR