2-Methylnaphtho[1,2-d]thiazole 98%

Supplier: Ambeed
Warning

A177552-10G A177552-1G
76998-424EA 50 USD
76998-424 76998-426
2-Methylnaphtho[1,2-d]thiazole 98%
2-Methylnaphtho[1,2-d]thiazole

Precaución: for Research Use Only

Formula: C₁₂H₉NS
MDL Number: MFCD00004951
CAS Number: 2682-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR