2-Bromo-6-fluoropyridine 98%

Supplier: Ambeed
Warning

A177502-100G A177502-10G A177502-25G A177502-500G A177502-5G
76998-414EA 99 USD
76998-414 76998-416 76998-418 76998-420 76998-422
2-Bromo-6-fluoropyridine 98%
2-Bromo-6-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrFN
MDL Number: MFCD03095258
CAS Number: 144100-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR