(4-Chloropyridin-2-yl)methanamine 98%

Supplier: Ambeed
Warning

A177452-100MG A177452-1G A177452-250MG A177452-5G
77011-828EA 10 USD
77011-828 77567-612 77890-133 77890-134
(4-Chloropyridin-2-yl)methanamine 98%
(4-Chloropyridin-2-yl)methanamine

Precaución: For research use only.

Formula: C₆H₇ClN₂
MDL Number: MFCD07374904
CAS Number: 180748-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR