5-(Benzyloxy)-1H-indole-3-carbonitrile 95%

Supplier: Ambeed
Warning

A177431-1G A177431-5G
77011-824EA 175.05 USD
77011-824 77011-826
5-(Benzyloxy)-1H-indole-3-carbonitrile 95%
5-(Benzyloxy)-1H-indole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂O
MW: 248.27 g/mol
MDL Number: MFCD13193421
CAS Number: 194490-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR