5,6-Diphenylpyrazin-2-ol 97%

Supplier: Ambeed
Warning

A177346-100G A177346-10G A177346-1G A177346-25G A177346-5G
77029-302EA 366.02 USD
77029-302 77029-304 77029-306 77029-308 77029-310
5,6-Diphenylpyrazin-2-ol 97%
5,6-Diphenylpyrazin-2-ol

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂O
MW: 248.27 g/mol
MDL Number: MFCD00234902
CAS Number: 18591-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR