2-Amino-3-bromo-5-chloropyridine 98%
Supplier: Ambeed
|
Warning
|
A177197-100G
A177197-10G
A177197-25G
A177197-500G
A177197-5G
76998-404EA
41
USD
76998-404
76998-406
76998-408
76998-410
76998-412
2-Amino-3-bromo-5-chloropyridine 98%
2-Amino-3-bromo-5-chloropyridine
Precaución: for Research Use Only
Formula:
C₅H₄BrClN₂ |
MDL Number:
MFCD00955627 CAS Number: 26163-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...