2-Iodo-5-methylthiophene 98%

Supplier: Ambeed
Warning

A176988-5G A176988-25G
76998-402EA 35 USD
76998-402 77881-345
2-Iodo-5-methylthiophene 98%
2-Iodo-5-methylthiophene

Precaución: for Research Use Only

Formula: C₅H₅IS
MDL Number: MFCD00022492
CAS Number: 16494-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR