tert-Butyl 3-acetylpyrrolidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A176932-100MG A176932-250MG A176932-1G A176932-5G
77029-280EA 45.62 USD
77029-280 77029-282 77593-126 77600-432
tert-Butyl 3-acetylpyrrolidine-1-carboxylate 98%
tert-Butyl 3-acetylpyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
MDL Number: MFCD11865235
CAS Number: 858643-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR