Nɑ-(tert-Butoxycarbonyl)-D-lysine 97%

Supplier: Ambeed
Warning

Synonyms: Nɑ-Boc-D-lysine, Boc-D-Lys-OH

A176922-1G A176922-25G A176922-5G
76998-396EA 10.61 USD
76998-396 76998-398 76998-400
Nɑ-(tert-Butoxycarbonyl)-D-lysine 97%
Nalpha-(tert-Butoxycarbonyl)-D-lysine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₄
MW: 246.31 g/mol
MDL Number: MFCD00076956
CAS Number: 106719-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR