2-Chloro-5-nitropyridine 95%

Supplier: Ambeed
Warning

A176909-100G A176909-10G A176909-25G A176909-500G A176909-5G A176909-1000G
76985-640EA 24.41 USD
76985-640 76985-642 76985-644 76985-646 76985-648 77683-416
2-Chloro-5-nitropyridine 95%
2-Chloro-5-nitropyridine

Precaución: For research use only.

Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
MDL Number: MFCD00006240
CAS Number: 4548-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR