2-Chloro-3-pyridineboronic acid 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-pyridinylboronic acid, (2-Chloropyridin-3-yl)boronic acid, 2-Chloropyridine-3-boronic acid

A176642-10G A176642-1G A176642-25G A176642-5G A176642-100G
77011-794EA 26 USD
77011-794 77011-796 77011-798 77011-800 77683-415
2-Chloro-3-pyridineboronic acid 98%, Ambeed.Inc
2-Chloro-3-pyridineboronic acid

Precaución: For research use only.

Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD03094997
CAS Number: 381248-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR