3-(Aminoethyl)benzo[b]furan 96%

Supplier: Ambeed
Warning

Synonyms: 2-(1-Benzofuran-3-yl)ethylamine, 2-(Benzo[b]furan-3-yl)ethylamine, 2-(Benzofuran-3-yl)ethanamine, 3-(Aminoethyl)-1-benzofuran

A176453-100MG A176453-1G A176453-250MG
77029-274EA 51.99 USD
77029-274 77029-276 77029-278
3-(Aminoethyl)benzo[b]furan 96%
3-(Aminoethyl)benzo[b]furan

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
MDL Number: MFCD08271905
CAS Number: 27404-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR