Bicalutamide 99%

Supplier: Ambeed
Warning

Synonyms: N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-((4-fluorophenyl)sulphonyl)-2-hydroxy-2-methylpropanamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulphonyl]-2-hydroxy-2-methylpropionamide

A176397-100MG A176397-1G A176397-250MG A176397-50MG A176397-5G
77029-264EA 6.37 USD
77029-264 77029-266 77029-268 77029-270 77029-272
Bicalutamide 99%
Bicalutamide

Precaución: for Research Use Only

Formula: C₁₈H₁₄F₄N₂O₄S
MW: 430.37 g/mol
MDL Number: MFCD00869971
CAS Number: 90357-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR