3-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol

A176243-100MG A176243-1G A176243-250MG
77011-778EA 10 USD
77011-778 77011-780 77011-782
3-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester 95%
3-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MDL Number: MFCD18732617
CAS Number: 1082066-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR