1,3-Dichloro-5-(3,3,3-trifluoroprop-1-en-2-yl)benzene 98% stabilized, Ambeed.Inc

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A176218-1G A176218-250MG A176218-5G A176218-25G
77011-772EA 53.05 USD
77011-772 77011-774 77011-776 77683-414
1,3-Dichloro-5-(3,3,3-trifluoroprop-1-en-2-yl)benzene 98% stabilized, Ambeed.Inc
1,3-Dichloro-5-(3,3,3-trifluoroprop-1-en-2-yl)benzene

Precaución: For research use only.

Formula: C₉H₅Cl₂F₃
MW: 241.03 g/mol
MDL Number: MFCD24387012
CAS Number: 864725-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR