2,4-Dichloro-5,6,7,8-tetrahydroquinazoline 97%

Supplier: Ambeed
Warning

A176071-10G A176071-1G A176071-250MG A176071-25G A176071-5G
77029-252EA 377.69 USD
77029-252 77029-254 77029-256 77029-258 77029-260
2,4-Dichloro-5,6,7,8-tetrahydroquinazoline 97%
2,4-Dichloro-5,6,7,8-tetrahydroquinazoline

Precaución: for Research Use Only

Formula: C₈H₈Cl₂N₂
MW: 203.0700 g/mol
MDL Number: MFCD12400774
CAS Number: 1127-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR