((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphonic acid 99+%

Supplier: Ambeed
Danger

Synonyms: Adefovir

A176069-5G A176069-25G
76998-366EA 10 USD
76998-366 77671-498
((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphonic acid 99+%
((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphonic acid

Precaución: for Research Use Only

Formula: C₈H₁₂N₅O₄P
MW: 273.19 g/mol
MDL Number: MFCD00866943
CAS Number: 106941-25-7
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR