3-Chloro-5-fluorobenzoic acid 95%

Supplier: Ambeed
Warning

A175779-100G A175779-1G A175779-25G A175779-5G
76998-358EA 299.18 USD
76998-358 76998-360 76998-362 76998-364
3-Chloro-5-fluorobenzoic acid 95%
3-Chloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
MDL Number: MFCD00973904
CAS Number: 25026-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR