Boc-L-β-homoleucine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-β-HoLeu-OH, (S)-3-((tert-Butoxycarbonyl)amino)-5-methylhexanoic acid

A175642-1G A175642-250MG
76998-354EA 76.39 USD
76998-354 76998-356
Boc-L-β-homoleucine 98%
Boc-L-beta-homoleucine

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₄
MW: 245.32 g/mol
MDL Number: MFCD02101665
CAS Number: 132549-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR