6-Oxo-1,6-dihydropyrimidine-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A175606-100MG A175606-10G A175606-1G A175606-250MG A175606-5G
77029-240EA 11.67 USD
77029-240 77029-242 77029-244 77029-246 77029-248
6-Oxo-1,6-dihydropyrimidine-5-carboxylic acid 97%
6-Oxo-1,6-dihydropyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄N₂O₃
MW: 140.1000 g/mol
MDL Number: MFCD08692965
CAS Number: 65754-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR